Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hyaluronic Antibody, Novus Biologicals™
SDP

Catalog No. NB017505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017505 0.1 mL
NB10062665 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017505 Supplier Novus Biologicals Supplier No. NB100626650.1ML
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Hyaluronic Polyclonal specifically detects Hyaluronic in Human samples. It is validated for ELISA.

Specifications

Antigen Hyaluronic
Applications ELISA
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:100-1:2000
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Host Species Sheep
Immunogen Hyaluronic acid from human umbilical cord
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62665 is specific for Hyaluronic acid, a mucopolysaccharide which is an integral part of the gel-like substance of animal connective tissue that serves as a lubricanting and protective agent.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.