Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Bovine Growth Serum

Catalog No. SH3054102 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated (25 to 40 doses)
None
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH3054102 100 mL None
SH305410340 500 mL Irradiated (25 to 40 doses)
SH3054102HI 100 mL Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3054102 Supplier Cytiva Supplier No. SH30541.02

A cost-effective alternative to Fetal Bovine Serum (FBS).

TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Description 100 mL
Product Type Bovine Growth Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing None
Tested For Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL
Mycoplasma Detected Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.