Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Bovine Serum Albumin (BSA)

Catalog No. p-7058032 Shop All Cytiva Products
Click to view available options
Quantity:
10 g
100 g

Catalog No. SH3057401

Supplier: Cytiva SH30574.01

Support cell culture growth with HyClone™ Bovine Serum Albumin (BSA).

  • pH 7.0
  • Lyophilized powder
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Packaged in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage 2° to 8° C
Form Lyophilized Powder
Product Type BSA
Sterility Non-sterile
Endotoxin Level ≤ 10 EU/mg
pH 6.5 to 7.5
Purity or Quality Grade Cell Culture Grade
Quantity 10 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.