Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (New Zealand Origin)

Catalog No. p-7145853 Shop All Cytiva Products
Click to view available options
Quantity:
500 mL
Processing:
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses

Enhance growth of cells with New Zealand sourced Characterized FBS.

  • Carefully collected, processed, and filtered in our own New Zealand facility
  • The great expanse of the Pacific ocean has protected New Zealand’s islands from many outside influences, both geographically and biologically.
  • Greater security against bovine disease, since New Zealand has the fewest reported bovine diseases in the world
  • Filtered through three sequential 100 nm (0.1 μm) pore size-rated filters
  • Before dispensing, each lot of serum is pooled to ensure uniformity and consistency between bottles using true pool technology
  • TRUSTED_SUSTAINABILITY

    Specifications

    Additional Assays Progesterone,Chloride,Insulin,Inorganic Phosphorus,PTH,Glucose,Cholesterol,Copper,HDL,Magnesium,LDL,Selenium,Triglycerides,IgG,Gamma Globulin,pH,Phospholipids,Osmolality,Alkaline Phosphatase,Iron,Lactate Dehydrogenase,TIBC,SGPT,Percent Saturation,SGOT,B1,B2,B6,Niacin,Biotin,Retinol,Carotene,Tocopherol,B12,Folate,Vitamin C,1,25-dihydroxy vitamin D,Total Protein,Albumin,Blood Urea Nitrogen
    Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
    Quantity 500 mL
    Sterility Sterile
    Sterilization Method Sequential-filtered, 100 nm
    Grade Characterized
    Processing Irradiated, 25 to 40 doses
    Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
    Endotoxin Concentration ≤ 20 EU/mL
    Heat Inactivated No
    Hemoglobin Concentration ≤ 25 mg/dL
    Mycoplasma Detected Yes
    USDA Safety Testing Included USDA Inspected
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.