Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva HyClone™ Cosmic Calf™ Serum (U.S.)
Shop All Cytiva Products
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Heat Inactivated
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses
None
Description
- Age at time of collection: typically 16 - 22 weeks
- Iron-supplemented with additional growth promoting components
Specifications
Specifications
| Additional Assays | Insulin, Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG |
| Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. |
| Quantity | 500 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential-filtered, 100 nm, Gamma-irradiated |
| Processing | Irradiated, 25 to 40 doses |
| Tested For | Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
| Heat Inactivated | No |
| Iron Supplemented | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction