Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva HyClone™ Donor Equine Serum (U.S.)
Shop All Cytiva Products
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
None
Description
- Format: Liquid

Specifications
Specifications
Additional Assays | Albumin, Alkaline Phosphatase, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, Inorganic Phosphorus, Iron, Lactate Dehydrogenase, Osmolality, Percent Saturation, pH, Potassium, SGOT, SGPT, Sodium, TIBC, Total Bilirubin, Total Protein |
Country of Origin | United States |
Form | Liquid |
Product Type | Equine Serum |
Quantity | 500 mL |
Sterilization Method | Sequential Triple-filtered, 100 nm |
Processing | Heat Inactivated |
Tested For | BVDV, Cytopathogenic Agents, Rabies Virus, Reovirus |
Heat Inactivated | Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction