Learn More
Cytiva HyClone™ Donor Equine Serum (U.S. origin)
Catalog No. p-7145855 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated (25 to 40 kGy)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Processing
SH300740340 500 mL Irradiated (25 to 40 kGy)
SH3007402HI 100 mL Heat Inactivated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH300740340 Supplier Cytiva Supplier No. SH30074.03IR2540
Only null left
Add to Cart
Add to Cart
Supplement cell culture growth with U.S. sourced, collected from selected donor herds that receive regular veterinary inspection and care. Complete traceability back to the original source.
  • Donor herds are fed an enriched diet to ensure proper nutrition and are fasted prior to bleeding to reduce lipid concentration
  • The horses are bled in an enclosed facility that is specially equipped for aseptic collection
  • Coggins tests for equine infectious anemia are routinely performed on each horse
  • Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles
  • TRUSTED_SUSTAINABILITY
    Specifications
    Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
    Quantity 500 mL
    Sterilization Method Sequential-filtered, 100 nm
    Processing Irradiated (25 to 40 kGy)
    Tested For BVDV, Rabies Virus, Reovirus, Cytopathogenic Agents
    Heat Inactivated No