Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Hyclone™ Defined Fetal Bovine Serum (U.S.)

Catalog No. SH300700345 Shop All Cytiva Products
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Processing:
Heat Inactivated
Irradiated (25 to 40 doses)
Irradiated (30 to 45 doses)
None

Catalog No. SH300700345

Supplier: Cytiva SH30070.03IR3045

Only null left
Add to Cart
Add to Cart

Used as serum-supplement for the in vitro cell culture of eukaryotic cells

  • Maximize cell culture growth
  • Tested for endotoxin, hemoglobin, and additional biochemical assays
  • Filtered through serial 40 nm pore-size rated filters
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays 25 Dihydroxy Vitamin D, Albumin, Alkaline Phosphatase, B1
Certifications/Compliance Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Country of Origin United States
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 40 nm
Grade Defined
Processing Irradiated (30 to 45 doses)
Tested For Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
Endotoxin Concentration ≤ 10 EU/mL
Heat Inactivated No
Hemoglobin Concentration ≤ 10 mg/dL
Product Line HyClone
Mycoplasma Detected No
USDA Safety Testing Included USDA Inspected
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.