Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Iron-Supplemented Bovine Calf Serum, US Origin

Catalog No. p-7087594 Shop All Cytiva Products
Click to view available options
Quantity:
1000 mL
Processing:
HI, Irr - 25 to 40 doses
This item is not returnable. View return policy
This item is not returnable. View return policy

Contains additional iron that boosts serum transferrin levels to physiological levels

  • Supplemented with iron and growth-promoting factors
  • Age of animal at time of collection approximately 16-24 weeks
  • Virus panel testing according to 9 CFR 113.53
  • Low in antibodies and high in growth factors
  • Sourced in the USA with complete traceability back to original source
  • A cost-effective alternative to FBS
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Manufactured in cGMP (21 CFR 820) Compliant and ISO9001 Certified Facilities
Age 16 to 22 weeks
Country of Origin United States
Product Type Calf Bovine Serum
Quantity 1000 mL
Sterilization Method Sequential-filtered, 100 nm
Source Formula-Fed Veal Calves
Processing HI, Irr - 25 to 40 doses
Tested For Virus Panel Testing According to 9 CFR 113.53
Endotoxin Concentration <10 EU/mL
For Use With (Application) Supplementing Cell Culture Growth
Heat Inactivated Yes
Hemoglobin Concentration <10 mg/dL
Iron Supplemented Yes
Product Line HyClone™
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.