Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Iron-Supplemented Calf Serum, U.S. origin

Catalog No. SH3007202 Shop All Cytiva Products
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
Processing:
Heat Inactivated
Irradiated, 25 to 40 doses
Irradiated, 30 to 45 doses
None

A cost-effective alternative to Fetal Bovine Serum (FBS). Contains additional iron to boost serum transferrin to physiological levels.

HyClone Iron-Supplemented Bovine Calf Serum, US Origin is collected by venipuncture from formula-fed veal calves, which produce exceptionally high levels of transferrin. This serum is carefully supplemented with iron to load serum transferrin levels to physiological levels. After supplementation, the serum contains three to four times as much available iron and transferrin as fetal bovine serum (FBS) or equine serum.

TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG
Content And Storage Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C
Product Type Calf Bovine Serum
Quantity 100 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing None
Tested For Bluetongue,Bovine Adenovirus,Bovine Parvovirus,Bovine RSV,BVDV,Cytopathogenic Agents,Hemadsorbing Agents,Rabies Virus,Reovirus
Heat Inactivated No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.