Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum (New Zealand)
Shop All Cytiva Products
Click to view available options
Quantity:
1000 mL
Processing:
Heat Inactivated
None
Description
Specifications
Specifications
| Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG |
| Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. |
| Country of Origin | New Zealand |
| Product Type | Newborn Calf Bovine Serum |
| Quantity | 1000 mL |
| Sterility | Sterile |
| Sterilization Method | Sequential-filtered, 100 nm |
| Processing | None |
| Tested For | Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus |
| Heat Inactivated | No |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction