Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum, New Zealand Origin

Catalog No. sh3062602hi Shop All Cytiva Products
Click to view available options
Quantity:
500 mL

Enhances cell culture growth, contains three to four times as much available iron and transferrin as FBS or equine serum

  • Tested for endotoxin and hemoglobin levels
  • Growth promotion studies have demonstrated various product applications
  • Often outperforms FBS in many applications
  • Sourced from animals that are typically less than 10 days old
TRUSTED_SUSTAINABILITY

Specifications

Additional Assays Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, Glutamic Pyruvic Transaminase (SGTP), Glutamic Oxaloacetic Transaminase (SGOT), Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorous, Glucose, pH, Osmolality, Iron, Total Iron Binding Capacity (TIBC), Percent Saturation, and IgG
Age <10 days
Country of Origin New Zealand
Form Frozen
Product Type Newborn Calf Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Processing Iron Supplemented, Heat Inactivated
Tested For Fluorescent Antibody, Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine Respiratory Syncytial Virus, Bovine Viral Diarrhea Virus, Rabies, Reovirus, Cytopathogenic Agents (e.g., IBR), and Hemadsorbing agents (e.g., PI3)
Heat Inactivated Yes
Product Line HyClone™
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.