Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ L-Glutamine, Powder

Catalog No. p-7087566 Shop All Cytiva Products
Click to view available options
Quantity:
100 g

For supplementing serum- and protein-free media

  • Support the growth of cells that have high energy demands
  • Synthesize large amounts of proteins and nucleic acids
TRUSTED_SUSTAINABILITY

Specifications

Amino Acids L-Glutamine
Form Powder
Product Type L-Glutamine
For Use With (Application) Supplementing Serum- and Protein-Free Media
Product Line HyClone™
Quantity 100 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.