Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HyClone™ PF-CHO™ LS and PF-CHO MPS Media

Catalog No. sh3033302 Shop All Cytiva Products
Click to view available options
Quantity:
1000 mL
5 L
10 L
With Additives:
Glutamine, Poloxamer 188, Sodium Bicarbonate
None
Without Additives:
No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate
No Phenol Red, No HEPES
Product Type:
PF-CHO LS
PF-CHO MPS

Support growth of multiple CHO cell clones and production of a variety of recombinant proteins. HyClone™ PF CHO™ LS and PF CHO MPS Media is a protein-free media developed through Metabolic Pathway Design.

  • Designed for high cell yield and recombinant protein production
  • Allow for direct or sequential adaptation
  • Support growth in multiple cell culture systems
  • Manufactured from traceable components according to cGMP (21 CFR 820) guidelines
  • TRUSTED_SUSTAINABILITY

    Specifications

    Form Powder
    Product Type PF-CHO MPS
    Sterility Non-sterile
    With Additives None
    Without Additives No Glutamine, No Phenol Red, No Poloxamer 188, No HEPES, No Sodium Bicarbonate
    Quantity 10 L
    Certifications/Compliance cGMP
    Content And Storage 2° to 8°C, away from light
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.