Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Hydrated-Sponge, Sold by AquaPhoenix Scientific
Shop All AquaPhoenix Scientific Products

Click to view available options
Includes:
Gloves HS10BPW2G
Gloves HS10LET2G
Gloves HS10NB2G
Saturation Solution:
10 mL Buffered Peptone Water Broth
10 mL letheen broth
10 mL neutralizing buffer
Description
- Ready-to-use, pre-hydrated sponges for easy and convenient environmental and food sample collection
- Durable sponge withstands scrubbing
- Sponge bag with write-on label for ease in labeling samples
- Includes two decontaminated gloves for safety and convenience
Specifications
Specifications
| Product Type | Hydrated Sponge |
| Length (English) | 3 in. |
| Length (Metric) | 7.6 cm |
| Width (English) | 1.5 in. |
| Width (Metric) | 4 cm |
| Packaging Quantity | 100/Cs. |
| Saturation Solution | 10 mL Buffered Peptone Water Broth |
| Includes | Gloves HS10BPW2G |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction