Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrazine Sulfate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. p-7157534 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
125 g
500 g
6 x 125 g
6 x 500 g
12 kg
50 kg
Packaging:
Drum
Poly Bottle
Poly Pail

H1018, 10034-93-2, H4N2·H2SO4

Spectrum™ Chemical Hydrazine Sulfate, Crystal, Reagent, ACS has been promoted to treat colon and rectal (colorectal) cancer, lung cancer, brain cancer (neuroblastoma), Hodgkin′s disease (lymph cancer), and other cancers. It is also used for the general weight loss and wasting (cachexia) associated with cancer. However, hydrazine sulfate is not approved for use with cancer patients in the United States. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 10034-93-2
Molecular Formula H6N2O4S
Molecular Weight (g/mol) 130.12
InChI Key ZGCHATBSUIJLRL-UHFFFAOYSA-N
IUPAC Name hydrazine; sulfuric acid
SMILES NN.OS(O)(=O)=O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 25 g
UN Number UN1759
Percent Purity 99%
Grade ACS
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.