Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrobromic Acid, purum p.a., Honeywell™

Catalog No. 6000696 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L

Catalog No. 60-006-96

Supplier: Honeywell Chemicals 187301L

Only null left
Add to Cart
Add to Cart

Purum p.a.,≥48%

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-10-6
Molecular Formula BrH
Molecular Weight (g/mol) 80.91
MDL Number MFCD00011323
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-N
Synonym hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
PubChem CID 260
ChEBI CHEBI:47266
IUPAC Name bromane
SMILES Br

Specifications

Density 1.49g/mL at 25°C
Vapor Pressure 8mmHg (25°C), 320psi (21.1°C)
Quantity 1 L
UN Number 1788
Beilstein 3587158
Formula Weight 80.91g/mol
Percent Purity ≥48%
Grade purum p.a.
Chemical Name or Material Hydrobromic Acid
Hazard Statement P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314-H335
EINECSNumber 233-113-0
RTECSNumber MW3850000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.