Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydrofluoric Acid, 48-52%, Technical, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
500 mL
1 gal.
6 x 500 mL
4 x 1 gal.
Description
Spectrum™ Chemical Hydrofluoric Acid, 48-52 Percent, Technical is a highly corrosive solution of hydrogen fluoride in water is capable of dissolving a wide range of materials including oxides and glass.. Used in a variety of fields including oil refining and production of fluorides it can also be used to dissolve rock samples before analysis.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 500 mL |
| UN Number | UN1790 |
| Grade | Technical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction