Learn More
Hydrofluoric acid, for analysis, 48 to 51% solution in water, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Melting Point | -35.0°C |
| Boiling Point | 105.0°C |
| Density | 1.1500g/mL |
| Heavy Metals (as Pb) | 0.5ppm max. |
| Packaging | Plastic bottle |
| Residue after Ignition | 0.0005% max. |
| Specific Gravity | 1.15 |
| Quantity | 25 mL |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement: Fatal in contact with skin. Causes severe skin burns and eye damage. Fatal if swallowed. Fatal if inhaled. |
| Precautionary Statement | GHS P Statement: Wear face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 231-634-8 |
| RTECSNumber | MW7875000 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.