Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrofluoric acid, Environmental Grade Plus, 47-51%, Thermo Scientific Chemicals

Catalog No. AA38746AP
Click to view available options
Quantity:
500 mL

Catalog No. AA38746AP

Supplier: Thermo Scientific Chemicals 038746AP

Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-39-3
Molecular Formula FH
Molecular Weight (g/mol) 20.01
MDL Number MFCD00011346
InChI Key KRHYYFGTRYWZRS-UHFFFAOYSA-N
Synonym hydrofluoric acid, hydrogen fluoride, fluorhydric acid, hydrofluoride, rubigine, fluorwasserstoff, hydrogen-fluoride, antisal 2b, fluorowodor polish, caswell no. 484
PubChem CID 14917
ChEBI CHEBI:29228
SMILES F

Specifications

Color Colorless
Boiling Point 112°C
Density 1.16 g/mL
Odor Acrid
Quantity 500 mL
Physical Form Liquid, Double Sub-Boiling Quartz Distillation
UN Number UN1790
Merck Index 14,4790
IUPAC Name fluorane
Formula Weight 20.01
Percent Purity 47 to 51%
Grade Environmental Plus
Chemical Name or Material Hydrofluoric acid
Show More Show Less
Hazard Category H290-H300+H310+H330-H314-H335
Hazard Statement GHS H Statement
H300-H310-H330-H314
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement P234-P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDROFLUORIC ACID
EINECSNumber 231-634-8
RTECSNumber MW7875000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.