Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastec Corporation™ Hydrogen Cyanide Detector Tube
SDP

Catalog No. 17532383 Shop All Nextteq Products
Change view
Click to view available options
Detection Limits:
0.2 to 10 ppm
17 to 2400 ppm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Limits
17-532-383 17 to 2400 ppm
17-532-384 0.2 to 10 ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-532-383 Supplier Gastec Corporation™ Supplier No. 12M
Add to Cart
Add to Cart

Hydrogen Cyanide Detector Tube for use with Gastec™ Hand Pump.

  • Choose from a variety of detection limits

Specifications

Detection Limits 17 to 2400 ppm
Components Measured Hydrogen Cyanide
For Use With (Equipment) Gastec Hand Pump
Type Gas Detection Tube
Quantity 1 Tube
Sample Type Gas
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.