Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gastec Corporation™ Hydrogen Cyanide Detector Tube
SDP

Hydrogen Cyanide Detector Tube for use with Gastec™ Hand Pump.

$112.90

Specifications

Components Measured Hydrogen Cyanide
For Use With (Equipment) Gastec Hand Pump
Type Gas Detection Tube
Quantity 1 Tube
Sample Type Gas
View More Specs

Products 2
Catalog Number Mfr. No. Detection Limits Price Quantity & Availability  
Catalog Number Mfr. No. Detection Limits Price Quantity & Availability  
17-532-384
SDP
View Documents
Gastec Corporation™
12LL
0.2 to 10 ppm
Each for $112.90
Only null left
Add to Cart
 
17-532-383
SDP
View Documents
Gastec Corporation™
12M
17 to 2400 ppm
Each for $112.90
Only null left
Add to Cart
 
Description

Description

  • Choose from a variety of detection limits
Specifications

Specifications

Hydrogen Cyanide
Gas Detection Tube
Gas
Gastec Hand Pump
1 Tube
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.