Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Potassium ATPase Beta Antibody (2G11), Novus Biologicals™
SDP

Catalog No. NB300583 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300583 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300583 Supplier Novus Biologicals Supplier No. NB300583
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 5 publications

Hydrogen Potassium ATPase Beta Monoclonal specifically detects Hydrogen Potassium ATPase Beta in Human, Mouse, Rat, Porcine, Bovine, Canine, Ferret, Rabbit samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot.

Specifications

Antigen Hydrogen Potassium ATPase Beta
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 2G11
Conjugate Unconjugated
Dilution Western Blot 1:4000, Flow Cytometry 1:50, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:2000, Immunohistochemistry-Frozen 1:2000, Dot Blot 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Gene Alias ATP6B, ATPase, H+/K+ exchanging, beta polypeptide, ATPase, H+/K+ transporting, beta polypeptide, Gastric H(+)/K(+) ATPase subunit beta, gastric H+/K+ ATPase beta subunit, gastric hydrogen-potassium ATPase, beta, potassium-transporting ATPase beta chain, potassium-transporting ATPase subunit beta, Proton pump beta chain
Gene Symbols ATP4B
Host Species Mouse
Immunogen Purified 34 kDa core peptide from deglycosylated hog gastric microsomes.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 496
Test Specificity Detects the beta-subunit of hydrogen/potassium ATPase.
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Ferret, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.