Learn More
Hydrogen tetrachloroaurate(III) trihydrate, ACS, 99.99% (metals basis), Au 49.0% min
Description
It is used for microanalysis of rubidium, cesium and in the determination of alkaloids. Usually used as a harbinger for the preparation of gold (Au) nanoparticles. It is used in the control rods of nuclear reactors and in the manufacturing of electric light filaments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Physical Form | Crystalline |
| Quantity | 1 g |
| Assay Percent Range | 99.99% (metals basis) |
| Solubility Information | Soluble in water. Insoluble in ether. |
| Formula Weight | 393.83 (339.79 Anhydrous) |
| Grade | ACS Reagent |
| Sensitivity | Hygroscopic; Light sensitive |
| Chemical Name or Material | Hydrogen tetrachloroaurate trihydrate, For analysis ACS |
Safety and Handling
| Hazard Category | H302-H314-H317-H335 |
| Precautionary Statement | P260-P264b-P270-P271-P272-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 240-948-4 |
| RTECSNumber | MD5428000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.