Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydroxylysine kinase Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB042102 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB042102 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB042102 Supplier Novus Biologicals Supplier No. NBP297137H

Rabbit Polyclonal Antibody

Hydroxylysine kinase Polyclonal specifically detects Hydroxylysine kinase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Hydroxylysine kinase
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias 5-hydroxy-L-lysine kinase, 5-hydroxylysine kinase, AGPHD1, aminoglycoside phosphotransferase domain-containing protein 1, hydroxylysine kinase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Hydroxylysine kinase fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123688
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.