Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

hydroxysteroid (17-beta) dehydrogenase 11 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB325906 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325906 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325906 Supplier Novus Biologicals Supplier No. NBP299361APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

hydroxysteroid (17-beta) dehydrogenase 11 Polyclonal antibody specifically detects hydroxysteroid (17-beta) dehydrogenase 11 in Human samples. It is validated for Western Blot

Specifications

Antigen hydroxysteroid (17-beta) dehydrogenase 11
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias 17betaHSD11, 17betaHSDXI, 17-beta-hydroxysteroid dehydrogenase 11, 17-beta-hydroxysteroid dehydrogenase type XI, 17-beta-hydroxysteroid dehydrogenase XI, 17bHSD11, CTCL-associated antigen HD-CL-03, dehydrogenase/reductase (SDR family) member 8, DHRS8, EC 1.1.1, EC 1.1.1.62, hydroxysteroid (17-beta) dehydrogenase 11, member 2,17-beta-HSD 11, PAN1BRETSDR2,17-BETA-HSD11, retSDR2, RetSDR2,17-BETA-HSDXI, SDR16C2,17BHSD11, T-cell lymphoma-associated antigen HD-CL-03,17-beta-HSD XI
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human hydroxysteroid (17-beta) dehydrogenase 11.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 51170
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.