Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hydroxyurea, USP, 97-103%, Spectrum™ Chemical

Click to view available options
Quantity:
5 g
25 g
50 g
100 g
500 g
12 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical Hydroxyurea, USP is an antineoplastic drug used in myeloproliferative disorders. It has also been considered useful in treating painful attacks in sickle-cell disease and diseases like AIDS. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 100% |
Ignition Residue | 0.5% |
Loss on Drying | 1% |
Packaging | Poly Pail |
Quantity | 12 kg |
Percent Purity | 97 to 103% |
Grade | USP |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction