Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-500 Medium-Duty Industrial Gloves

Catalog No. p-8005290 Shop All Ansell Products
Click to view available options
Size:
8
9
This item is not returnable. View return policy
This item is not returnable. View return policy

Industrial gloves offer comfort and a sure grip. Ideal for dry or slightly oil applications where workers need light cut protection.

  • Ideal for assembly and maintenance operations involving handling of sharp-edged and pointed tools and materials
  • Cut-resistant stretch Kevlar™ liner provides comfortable fit and protection from nuisance nicks and abrasion
  • Black foam nitrile coating on palm and fingertips channels oil away from glove surface
  • Extended cuff for extra cut protection around wrist
  • Yellow
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009; Category III
Construction Knit
Coating Material Nitrile Foam
Color Black, Yellow
Cut Resistance Rating ANSI A2
Latex Free Yes
Product Type Cut-resistant Gloves
Size 8
EN388 Standard EN 388:3231B, EN 388:3231
For Use With (Application) Automotive, machinery and equipment, metal fabrication
Glove Cuff Style Knit
Liner Material Nylon, Lycra™, Aramid Fiber
Model 11-500 8 205576
Series HyFlex™ 11-500
Length (English) 7.8 in.
Length (Metric) 19.8 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.