Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 11-812 Foam Nitrile Coated Tearable Gloves

Catalog No. 19500755 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
This item is not returnable. View return policy
This item is not returnable. View return policy

A uniquely knit design allows the glove to easily tear at multiple high risk areas, significantly reducing the risk of hand injury if the glove becomes entangled in a rotating tool.

  • Ultra-thin assembly gloves designed to easily tear apart at multiple high risks areas
  • Foam nitrile coating provides extreme tactility when a high degree of precision is required
  • ZONZ Comfort Fit Technology keeps hands cool and comfortable
  • Gloves will not directly protect workers from injury caused by contact with a sharp or high-speed rotating machine
  • Discard gloves that have become entangled in a rotating machine, whether or not it has been torn
  • Recommended for rotating tools with a minimum torque of 10 Nm, and speeds over 200 RPM
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 2
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Chlorinated No
Construction Knit
Coating Material Nitrile Foam
Color White
Exterior Finish No Texture
Hand Specific
Material Nylon, Spandex
Product Type Seamless Knit Gloves
Size 9
EN388 Standard 2010A
For Use With (Application) Automotive Assembly, Machinery/Equipment Operation, Metal Fabrication, and Warehousing
Gender Unisex
Glove Cuff Style Knit
Grip Design Rough
Series HyFlex™ 11-812
Yarn Gauge 18
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.