Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell HyFlex™ 74-731 Knife-Hand Cut Protection Gloves

Catalog No. 19501024 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy

Catalog No. 19-501-024

Supplier: Ansell 284986

This item is not returnable. View return policy

Knife-Hand lightweight gloves are manufactured using patented ZONZ™ Comfort Fit Technology to match the shape of the hand, and to reduce fatigue/risk of cut injury.

  • Dyneema™ Diamond Technology, providing enhanced comfort & protection in high-risk areas
  • Specially designed to meet the individual needs associated with knife-hand and off-hand applications
  • Features Tuff Cuff II Technology to help retain cuff shape, keeping gloves secure and reducing worker injury
  • Compliant with FDA food handling requirements
  • Uses:
    • Food processing (beef, pork, lamb, poultry, seafood, fruits and vegetables)
  • TRUSTED_SUSTAINABILITY

    Specifications

    ANSI Abrasion Rating ANSI 4
    Certifications/Compliance FDA
    Certifications/Compliance (Europe) EN 420:2003, A1:2009
    Construction Knit
    Color Green, Gray
    Cut Resistance Rating ANSI A4
    Hand Specific
    Latex Free Yes
    Product Type Cut-resistant Gloves
    Size 8
    EN388 Standard EN 388:4X4XD, EN 388:454X
    For Use With (Application) Food processing
    Glove Cuff Style Tuff-Cuff™ II
    Liner Material Dyneema™ Diamond Technology, Polyester, Nylon, Spandex, Stainless Steel
    Model 284986
    Series HyFlex™ 74-731
    Length (English) 8.5 in.
    Length (Metric) 21.8 cm
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Your feedback has been submitted: Thank you for helping us improve our website.