Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYPE Antibody (OTI2A11), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB271681A48 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB271681A48 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB271681A48 Supplier Novus Biologicals Supplier No. NBP271681AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HYPE Monoclonal specifically detects HYPE in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen HYPE
Applications Western Blot
Classification Monoclonal
Clone OTI2A11
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AMPylator FICD, EC 2.7.7.n1, FIC domain containing, FIC domain-containing protein, fic S-phase protein cell division homolog, HIP-13, HIP13UNQ3041, huntingtin interacting protein 13, Huntingtin interacting protein E, huntingtin interactor protein E, Huntingtin yeast partner E, Huntingtin-interacting protein 13, Huntingtin-interacting protein E, HYPEadenosine monophosphate-protein transferase FICD, MGC5623
Gene Symbols FICD
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 61-265 of human HYPE (NP_009007) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 11153.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.