Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyperSep™ Polystyrene Lab Plates

Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.

Specifications

For Use With (Application) Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
Product Line HyperSep
Type 96-Well Plate
View More Specs

Products 5
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
03-250-555
View Documents
Thermo Scientific™
60110202
C8; Polystyrene N/A N/A N/A
03-250-556
View Documents
Thermo Scientific™
60110203
C4; Polystyrene N/A N/A N/A
03-250-557
View Documents
Thermo Scientific™
60110204
Hypercarb; Polystyrene N/A N/A N/A
03-250-558
View Documents
Thermo Scientific™
60110205
C18 plus Hypercarb; Polystyrene N/A N/A N/A
03-250-561
View Documents
Thermo Scientific™
60110208
SCX; Polystyrene N/A N/A N/A
Description

Description

Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polystyrene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include:

  • Tissue culture and separation of products
  • Concentration of small-scale samples
  • Sample clean-up
  • Collection of sample after chromatography

The convenient 96-well HyperSep Lab Plates can be processed manually or using a liquid-handling robot. They are not suitable for use with a vacuum.
Specifications

Specifications

Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
HyperSep
96-Well Plate
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.