Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Hypersil GOLD™ C4 Analytical HPLC Columns, 1.9μm

Catalog No. 25502054630
Change view
Click to view available options
Diameter (Metric):
1 mm
4.6 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
25-502-054630 4.6 mm
25-502-051030 1 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-502-054630 Supplier Thermo Scientific™ Supplier No. 25502054630

Achieve faster separations with excellent peak shapes for very hydrophobic analytes using Thermo Scientific™ Hypersil GOLD™ C4 Analytical HPLC columns.

Achieve faster separations with excellent peak shapes for very hydrophobic analytes using Thermo Scientific™ Hypersil GOLD™ C4 Analytical HPLC columns. Featuring a lower hydrophobicity than C18 or C8 columns, Hypersil GOLD C4 columns provide outstanding sensitivity and high efficiency.

Similar Selectivity, Less Retention Than C18 or C8 Columns
  • Successful retention and separation of very hydrophobic analytes with this short alkyl chain length, low hydrophobicity column
  • Ideal for analysis of hydrophobic polypeptides and proteins
Superior Performance
  • Highly symmetrical peak shapes
  • 1.9μm particle size columns improve speed and efficiency

About the Hypersil GOLD Family of HPLC Columns
  • Twelve different chemistries are available
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5μm particle sizes for all your separation needs
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4.6 mm
Flow Rate 2.5 mL/min
For Use With HPLC
Length (Metric) 50 mm
Max. Pressure 18130 psi (1250 bar)
Particle Size 1.9 μm
pH 2 to 8
Pore Size 175 Å
Surface Area 220 m²/g
Temperature 60°C
USP Type L26
Stationary Phase C4
Packing Material Silica
Carbon Load 5%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.