Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hypusine Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$240.00 - $598.00
Specifications
| Antigen | Hypusine |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Serum |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ABS1064I2MI
|
MilliporeSigma
ABS1064I25UL |
25 μL |
Each for $240.00
|
|
|||||
|
ABS1064I1MI
|
MilliporeSigma
ABS1064I100UL |
100 μL |
Each for $598.00
|
|
|||||
Description
Hypusine Polyclonal antibody specifically detects Hypusine in Human samples. It is validated for Western Blotting.Specifications
| Hypusine | |
| Polyclonal | |
| Serum | |
| Signaling | |
| N -4-amino-2-hydroxybutyl(lysine) | |
| IgG | |
| Unpurified | |
| This rabbit polyclonal antibody specifically detects hypusinated form of elF-5A. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Synthetic hypusine-containing peptide C-Ahx-STSKTG(hypusine)HGHAKV-amide. | |
| Primary | |
| Stable for 1 year at -20°C from date of receipt. | |
| ∼45 kDa observed (GFP-elf-5A). Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title