Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, Cy3
Shop All Mirus Bio LLC Products
Click to view available options
Quantity:
For 100 μg nucleic acid
For 5 μg nucleic acid
Description
Includes
Label IT™ Reagent, Label IT™ reconstitution solution, 10X Labeling Buffer and solutions
- Kits label any DNA or RNA template and are suitable for a wide range of applications
- One-step chemical method provides easy and consistent control of the labeling reactions
- Adjustable labeling density permits DNA and RNA to be optimally labeled for high sensitivity results
- Covalent mechanism provides permanent, nondestructive modification of nucleic acid residues
- Labels do not impact hybridization performance
Specifications
Specifications
Detection Method | Fluorescence |
Format | Kit |
Label or Dye | Cyanine3 |
Final Product Type | DNA (Labeled), RNA (Labeled) |
Labeling Target | DNA (General), Oligos, RNA (General) |
For Use With (Application) | Efficient, direct, nonenzymatic labeling of DNA and RNA |
Includes | Label IT™ Reagent, Label IT™ reconstitution solution, 10X Labeling Buffer and solutions |
Product Type | Label IT Cy3 Labeling Kit |
Quantity | For 5 μg nucleic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction