Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, Fluorescein

Catalog No. MIR3225 Shop All Mirus Bio LLC Products
Click to view available options
Quantity:
For 100 μg nucleic acid
For 25 μg nucleic acid

Includes

Label IT™ Reagent, Label IT™ reconstitution solution, 10X Labeling Buffer and solutions

Use the Label IT™ Nucleic Acid Labeling Kit, Fluorescein to efficiently and reproducibly attach marker molecules to DNA or RNA in a simple one-step reaction within minutes. Covalently attach labels without impacting hybridization performance.

  • Kits label any DNA or RNA template and are suitable for a wide range of applications.
  • One-step chemical method provides easy and consistent control of the labeling reactions.
  • Adjustable labeling density permits DNA and RNA to be optimally labeled for high sensitivity results.
  • Covalent mechanism provides permanent, nondestructive modification of nucleic acid residues.
  • Labels do not impact hybridization performance.

Specifications

Detection Method Fluorescence
Format Kit
Label or Dye FITC
Final Product Type DNA (Labeled), RNA (Labeled)
Labeling Target DNA (General), Oligos, RNA (General)
For Use With (Application) Efficient, direct, nonenzymatic labeling of DNA and RNA
Includes Label IT™ Reagent, Label IT™ reconstitution solution, 10X Labeling Buffer and solutions
Product Type Label IT Fluorescein Labeling Kit
Quantity For 25 μg nucleic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.