Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ambion™ mirVana™ miRNA Mimic

Catalog No. p-7075567
Encompass_Preferred
Click to view available options
Quantity:
250 nmol

Catalog No. 44-640-70

Supplier: Ambion™ 4464070

Only null left
Add to Cart
Add to Cart

Small, chemically modified double-stranded RNAs that mimic endogenous miRNAs and enable miRNA functional analysis by up-regulation of miRNA activity.

mirVana miRNA Mimics are designed to mimic endogenous microRNAs (miRNAs) and are excellent for studying miRNA function in various biological processes. These synthetic molecules are chemically modified to enhance stability and performance in both in vitro and in vivo applications.

mirVana miRNA Mimics are available for a wide range of miRNAs, providing researchers with a powerful tool to investigate gene regulation mechanisms.

Key features include:

  • High specificity and potency—designed to closely resemble natural miRNAs, helping ensure high specificity and potent gene regulation
  • Enhanced stability—chemical modifications enhance stability, making them suitable for long-term studies
  • Broad range of targets—available for a comprehensive list of miRNAs from miRBase v22, allowing for extensive research possibilities
  • Optimized for delivery—compatible with various transfection reagents and delivery systems, ensuring efficient uptake by cells
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Description mirVana™ Mimics and Inhibitors, 250nmol, Small, Chemically Modified Double-stranded RNAs that Mimic Endogenous miRNAs and Enable miRNA Functional Analysis by Up-regulation of miRNA Activity
Format 96-well Plate, 24-well Plate, 6-well Plate
RNAi Type miRNA
Final Product Type mirVana™ Mimics and Inhibitors
Quantity 250 nmol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.