Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cancer Diagnostics, Inc.™ i-Shields™ Eyeshields
Shop All Cancer Diagnostics Products

Click to view available options
Color:
Assorted
Clear
No. per Pack:
10/Pk.
100/Cs.
100/Pk.
25/Pk.
LABPAK Professional Pack (25 Lenses and 10 Frames)
Product Type:
Eyeshield frame
Eyeshield frame and lens
Eyeshield frames
Eyeshield lens
Description
Includes
Eyeshield FRAME only. (Lens is sold separately.)
Can be worn with a surgical mask.
- Ultra Lightweight for stress free/mark free extended wearing
- Reusable frame and disposable lenses provide side shield protection
- Distortion-free, optical grade lenses work well with most sizes of prescription eyewear and can be worn with a surgical mask
- Frames packaged in a variety of colors
- Lightweight recyclable components can be easily sterilized
Order Info
NOTE: Lenses and frames are ordered separately unless ordered in combo pack (such as 22-500-452).
Specifications
Specifications
| Color | Assorted |
| Product Type | Eyeshield frame |
| For Use With (Equipment) | most sizes of prescription eyewear and can be worn with a surgical mask |
| No. per Pack | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction