Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Taq Buffers (10X)
Click to view available options
Chemical Name or Material:
Taq Buffer with (NH4)2SO4
Taq Buffer with (NH4)2SO4 and MgCl2
Taq Buffer with KCl
Taq Buffer with KCl and MgCl2
Taq Buffer without Detergent
Description
Thermo Scientific 10X Taq Buffers are ready-to-use buffers for PCR using Taq DNA polymerases (both recombinant and native). They are available in different compositions and also without detergents.
10X Taq Buffer without Detergent
- 100 mM Tris-HCl (pH 8.8 at 25°C)
- 500 mM KCl
10X Taq Buffer with KCl and 15 mM MgCl2
- 100 mM Tris-HCl (pH 8.8 at 25°C)
- 500 mM KCl
- 15 mM MgCl2
- Detergent
10X Taq Buffer with (NH4)2SO4
- 750 mM Tris-HCl (pH 8.8 at 25°C)
- 200 mM (NH4)2SO4
- 0.1% (v/v) Tween 20
10X Taq Buffer with (NH4)2SO4 and 20 mM MgCl2
- 750 mM Tris-HCl (pH 8.8 at 25°C)
- 200 mM (NH4)2SO4
- 0.1% (v/v) Tween 20
- 20 mM MgCl2
10X Taq Buffer with KCl
- 100 mM Tris-HCl (pH 8.8 at 25°C)
- 500 mM KCl
- Detergent

Specifications
Specifications
Chemical Name or Material | Taq Buffer without Detergent |
Concentration | 10X |
For Use With (Application) | Standard PCR |
Physical Form | Liquid |
Quantity | 4 x 1.25 mL |
Includes | • 10X Taq Buffer without Detergent, 4 x 1.25 mL Store at -20°C. |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction