Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ iBET-BD2
GREENER_CHOICE

Catalog No. p-7222999
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective pan-BD2 inhibitor; orally bioavailable

iBET-BD2 is a potent and selective pan-BD2 inhibitor (IC50 values are 49, 98, 214 and 264 nM for BRD4BD2, BRD3BD2, BRDTBD2 and BRD2BD2, respectively). iBET-BD2 is selective for BD2 over BD1 (IC50 values are 10965, 36317, 70558, and >50119 ?M for BRD2BD1, BRD3BD1, BRD4BD1, and BRDTBD1, respectively) in a time-resolved FRET (TR-FRET) assay. iBET-BD2 (1 μM) inhibits IFN-γ-induced protein expression of MHC class I in K562 cells. iBET-BD2 reduces the production of anti-KLH IgM antibodies in mice immunized with keyhole limpet hemocyanin (KLH). iBET-BD2 exhibits immunomodulatory activity. iBET-BD2 is orally bioavailable.

Specifications

Chemical Name or Material 4-(Acetylamino)-3-fluoro-N-(trans-4-hydroxycyclohexyl)-5-[(1 S)-1-phenylethoxy]benzamide
CAS 2474876-09-8
Quantity 10 mg
Target Bromodomain Inhibitors
Molecular Formula C23H27FN2O4
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.