Learn More
Description
- Comes with a lid
- Available in MINI, Standard and Large
Specifications
Specifications
| Color | Clear |
| For Use With (Equipment) | Blot Washer System |
| Type | Large Blot Tray |
| Material | Polystyrene |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.