Learn More
Invitrogen™ iBind™ Flex Solution Kit

Description
• Optimized solutions with ideal viscosity for blocking, washing, and antibody dilution
• Long shelf life—store iBind Flex Solution Kit for 12 months at 4°C
• Better antigen-antibody interaction—solutions flow between the iBind Flex Card and membrane for maximum interaction between protein and antibody

Specifications
Specifications
Membrane Compatibility | Nitrocellulose, PVDF |
Detection Method | Colorimetric, Chromogenic, Chemiluminescence |
Product Line | iBind™ |
Product Type | Flex Solution Kit |
No. of Reactions | 10 midi blots or 20 mini blots |
Content And Storage | iBind Flex 5X buffer (400 mL) iBind Flex 100X additive (12 x 1.7 mL) Store at 2–8°C. |
Shipping Condition | Qualified for Ambient or Wet Ice |
For Use With (Equipment) | iBind Flex Western Device |
Quantity | 4 kits |
For Use With (Application) | Western Blotting |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.