Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ iBlot™ 3 Starter Kit, NC

Catalog No. A56728
Encompass_Preferred
Click to view available options
Warranty:
REX Extended Warranty, 2 years
Standard, 1-year warranty

The iBlot 3 Starter Kit includes the iBlot 3 Western Blot Transfer Device and two boxes of iBlot 3 Transfer Stacks, Midi, NC (nitrocellulose)—everything needed to perform a rapid dry-transfer of proteins from a gel to a membrane.

The iBlot 3 Starter Kit includes the iBlot 3 Western Blot Transfer Device and two boxes of iBlot 3 Transfer Stacks, Midi, NC (nitrocellulose)—everything needed to perform a rapid dry-transfer of proteins from a gel to a membrane.

iBlot 3 Western Blot Transfer Device

The iBlot 3 Western Blot Transfer Device is a dry transfer device that performs western blotting transfer efficiently and reliably in as few as three minutes and without the need for liquid buffers. The iBlot 3 system is compatible with both nitrocellulose and polyvinylidene difluoride (PVDF) membranes and offers equivalent or better performance compared with traditional wet-transfer methods in a fraction of the time. The iBlot 3 Western Blot Transfer Device is an integral part of the iBlot dry blotting system, which consists of the transfer device and consumable transfer stacks that contain the required buffers and transfer membrane (nitrocellulose or PVDF).

iBlot 3 Western Blot Transfer Device features include:

  • Complete protein transfer in as few as three minutes
  • High detection sensitivity and even, consistent protein transfer
  • Two independently controlled transfer stations capable of transferring up to two midi or four mini blots
  • Built-in adjustable cooling to ensure consistent transfer temperature
  • Pre-programed transfer methods optimized for low, high, and broad molecular weight ranges
  • Ability to create custom programs
  • A simple, user-friendly interface

How the iBlot dry blotting system works

Buffer ion reservoirs are incorporated into the gel matrix of transfer stacks instead of buffer tanks or soaked filter papers. The high density of ions in the gel matrix enables rapid protein transfer. During blotting, the copper anode does not generate oxygen gas as a result of water electrolysis, reducing blot distortion (conventional protein transfer techniques, including wet, semi-wet, and semi-dry, use inert electrodes that generate oxygen).

With the iBlot dry blotting system, transfer time is reduced by the shortened distance between electrodes, high field strength, and high current. Trapped air bubbles, often created during the manual preparation of the blotting sandwich layers, are easy to avoid due to our unique de-bubbling design that promotes even and complete transfer. With the iBlot 3 system, there is no need to prepare buffers or clean up after blotting. The total preparation and run time is normally less than ten minutes per blot.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • iBlot 3 Gel Transfer Device
• Western Blot Roller
• iBlot 3 Transfer Stacks, midi, nitrocellulose (2 boxes, 20 stacks)
• REX Extended Warranty

Store at room temperature.
For Use With (Application) Western Blot
For Use With (Equipment) iBlot 3 Transfer Stacks
Gel Compatibility Mini- or Midi-sized: Bis-Tris Gels, Tricine Gels, Tris-Acetate Gels, and Tris-Glycine Gels
Membrane Compatibility Nitrocellulose, PVDF
Mode of Transfer Dry
Product Line iBlot 3
Running Time ∼3 to 10 min.
Throughput 2 Midi or 4 Mini Gels
Type Starter Kit
Quantity 1 kit with warranty
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Warranty REX Extended Warranty, 2 years
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.