Learn More
Invitrogen™ iBlot™ Transfer Stack, PVDF, mini

Description
Includes
One box of iBlot Transfer Stacks containing 10 stacks
The iBlot Transfer Stacks are used to transfer proteins using the iBlot Gel Transfer Device. The iBlot Transfer Stacks are disposable stacks that come in three varieties for different applications: iBlotGel Transfer Stacks have integrated PVDF or nitrocellulose transfer membranes to perform dry blotting of proteins; iBlot DNA Transfer Stacks have an integrated nylon transfer membrane to perform dry blotting of DNA, and iBlot Western Detection Stacks are used to perform Western detection on previously blotted nitrocellulose or PVDF membranes. Each iBlot Transfer Stack contains a copper electrode and appropriate cathode and anode buffers in the gel matrix to allow fast, reliable transfer of proteins or DNA.
PVDF membrane
- 0.2μm pore size, low fluorescence
- Has higher binding capacity than nitrocellulose
- Preactivated and ready for use without any pretreatment with alcohols
- Compatible with commonly used detection methods such as staining, immunodetection, fluorescence, or radiolabeling
- Bind proteins due to hydrophobic interactions
- Protein binding capacity: 240μg/cm2
Transfer stack sizes
- The regular size (13 x 8.3cm) is used for all Novex midi-gel types (or equivalent) and E-PAGE™ 48 or 96 Gels
- The mini size (8 x 8cm) is used for all Novex mini-gels (or equivalent)
Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Length (Metric) | 8 cm |
| Width (Metric) | 8 cm |
| Quantity | 10 sets |
| Format | Transfer Stack |
| Material | PVDF |
| Pore Size | 0.2 μm |
| Content And Storage | 10 sets of transfer stacks in 1 box. Store at room temperature. |
| Shipping Condition | Room Temperature |
| Dimensions (LxW) | Mini, 8 cm x 8 cm |
| Product Line | iBlot |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.