Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IBRDC2 Antibody (OTI9H10), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP272470G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP272470G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP272470G Supplier Novus Biologicals Supplier No. NBP272470G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IBRDC2 Monoclonal specifically detects IBRDC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen IBRDC2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI9H10
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias bA528A10.3, E3 ubiquitin-protein ligase RNF144B, EC 6.3.2, EC 6.3.2.-, IBR domain containing 2, IBR domain-containing protein 2, IBRDC2, KIAA0161, p53-inducible RING finger protein, P53RFP, PIR2, ring finger protein 144BMGC71786
Gene Symbols RNF144B
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-256 of human IBRDC2 (NP_877434) produced in E. coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 255488.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.