Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ibrutinib
GREENER_CHOICE

Catalog No. p-7161633
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective BTK inhibitor (IC50= 0.5 nM)

Ibrutinib is a potent and selective BTK inhibitor (IC50= 0.5 nM). Selective for BTK against a screening panel of kinase enzymes. Inhibits autophosphorylation of BTK, phosphorylation of PLCγ, and phosphorylation of ERK (IC50 of 11 nM, 29 nM and 13 nM, respectively). Also inhibits ErbB4 (IC50 = 0.25-3.4 nM). Induces cytotoxicity of chronic lymphocytic leukemia (CLL) cells. Reduces clinical arthritis scores in a mouse model.

Specifications

Chemical Name or Material 1-[(3R)-3-[4-Amino-3-(4-phenoxyphenyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-piperidinyl]-2-propen-1-one
CAS 936563-96-1
Synonym Agammaglobulinaemia tyrosine kinase, AGMX1, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, tyrosine-protein kinase
Quantity 10 mg
Molecular Formula C25H24N6O2
Formula Weight 440.5
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.