Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ibuprofen Antibody, Novus Biologicals™
SDP

Catalog No. NB036468 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036468 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036468 Supplier Novus Biologicals Supplier No. NBP288849
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Ibuprofen Polyclonal specifically detects Ibuprofen in Non-species specific samples. It is validated for ELISA, Immunoaffinity Purification.

Specifications

Antigen Ibuprofen
Applications ELISA, Affinity Purification
Classification Polyclonal
Concentration 0.8 mg/ml
Conjugate Unconjugated
Dilution ELISA 1:200, Immunoaffinity Purification
Formulation Whole antiserum
Gene Alias 2-(4-ISOBUTYLPHENYL)PROPIONIC ACID, 4-Isobutylhydratropic acid
Host Species Rabbit
Immunogen BSA-Ibuprofen derivative conjugate
Purification Method Caprylic acid precipitation
Quantity 0.5 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.