Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ICA 110381
GREENER_CHOICE

Catalog No. 495010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-501-0 10 mg
49-505-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 49-501-0 Supplier Tocris Bioscience™ Supplier No. 4950/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

KV7.2/7.3 activator; displays anticonvulsant properties

KV7.2/7.3 activator (EC50 = 0.38 μM). Decreases neuronal excitability in CA1 hippocampal neurons. Exhibits anticonvulsive properties in amygdala-kindled rats, a model for complex partial seizures.

Chemical Identifiers

CAS 325457-99-6
Molecular Formula C12H8Cl2N2O

Specifications

Quantity 10 mg
Formula Weight 267.11
Purity >98%
Molecular Weight (g/mol) 267.11
Chemical Name or Material ICA 110381
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.