Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ICA1 Monoclonal Antibody (C7)
Mouse Monoclonal Antibody
$243.00 - $552.50
Specifications
Antigen | ICA1 |
---|---|
Clone | C7 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541622
|
Invitrogen™
MA541622 |
200 μL |
Each for $552.50
|
|
|||||
PIMA543699
|
Invitrogen™
MA543699 |
20 μL |
Each for $243.00
|
|
|||||
Description
ICA1 Monoclonal Antibody for Western Blot
This gene encodes a protein with an arfaptin homology domain that is found both in the cytosol and as membrane-bound form on the Golgi complex and immature secretory granules. This protein is believed to be an autoantigen in insulin-dependent diabetes mellitus and primary Sjogren's syndrome. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized.Specifications
ICA1 | |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q05084 | |
3382 | |
Recombinant protein Islet Cell Autoantigen 1. The antigen corresponds to amino acid range 1-258 of the target protein. | |
Antibody |
C7 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2b κ | |
Human | |
ICA1 | |
69 kDa islet cell autoantigen; 69kDa; diabetes mellitus type I autoantigen; ica1; ica1 {ECO:0000312; ICA69; ICAp69; Islet cell autoantigen 1; islet cell autoantigen 1 isoform; islet cell autoantigen 1, 69 kDa; islet cell autoantigen 1, 69kDa; islet cell autoantigen p69; p69; RGD:621465}; testicular tissue protein Li 162; Unknown (protein for MGC:134001); zgc:92566 | |
ICA1 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title