Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ICAM-1/CD54 Antibody (101), Alexa Fluor™ 700
SDP

Catalog No. NB040115 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB040115 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB040115 Supplier Novus Biologicals Supplier No. NBP290587AF700

Rabbit Monoclonal Antibody

ICAM-1/CD54 Monoclonal specifically detects ICAM-1/CD54 in Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Sandwich ELISA Detection.

Specifications

Antigen ICAM-1/CD54
Applications Immunofluorescence, ELISA
Classification Monoclonal
Clone 101
Conjugate Alexa Fluor 700
Dilution Immunocytochemistry/Immunofluorescence, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias BB2, CD54, CD54 antigen, cell surface glycoprotein P3.58, human rhinovirus receptor, ICAM-1, intercellular adhesion molecule 1, intercellular adhesion molecule 1 (CD54), human rhinovirus receptor, Major group rhinovirus receptor, P3.58
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse ICAM-1/CD54 (NP_034623.1; Met1-Asn485).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Cell Biology, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3383
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.